Tris(2-methoxyphenyl)phosphine ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Tris(o-methoxyphenyl)phosphine
15-7950-1G
15-7950-5G
100201-508EA
73.6
USD
100201-508
100205-920
Tris(2-methoxyphenyl)phosphine ≥98%
Tris(2-methoxyphenyl)phosphine
Phosphine
Formula:
C₂₁H₂₁O₃P Melting Pt: 203…205 °C |
MDL Number:
MFCD00014892 CAS Number: 4731-65-1 EINECS: 225-235-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...