Tris(dimethylamino)phosphine ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

15-7800-1G 15-7800-5G
100197-426EA 43.88 USD
100197-426 100211-028
Tris(dimethylamino)phosphine ≥98%
Tris(dimethylamino)phosphine

Phosphine

Formula: C₆H₁₈N₃P
Boiling Pt: 49…51 °C (12 mmHg)
Melting Pt: –44 °C
Density: 0.8962 g/cm³ (20 °C)
Flash Pt: 98 °F
MDL Number: MFCD00008301
CAS Number: 1608-26-0
EINECS: 216-534-4
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR