Triphenyl phosphite ≥97%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
15-6850-500G
15-6850-2000G
100206-440EA
100.49
USD
100206-440
100206-438
Triphenyl phosphite ≥97%
Triphenyl phosphite
Formula:
C₁₈H₁₅O₃P MW: 310.29 g/mol Boiling Pt: 360 °C (1013 hPa) Melting Pt: 22 - 25 °C Density: 1.19 g/cm³ (20 °C) Flash Pt: 188 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003032 CAS Number: 101-02-0 EINECS: 202-908-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...