Triphenylphosphine oxide ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

15-6750-25G 15-6750-100G
100209-742EA 65.11 USD
100209-742 100208-842
Triphenylphosphine oxide ≥98%
Triphenylphosphine oxide
Formula: C₁₈H₁₅OP
MW: 278.29 g/mol
Boiling Pt: ∼360 °C (1013 hPa)
Melting Pt: 151…154 °C
Density: 1.2 g/cm³ (20 °C)
Flash Pt: 356 °F
MDL Number: MFCD00002080
CAS Number: 791-28-6
EINECS: 212-338-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR