Trimethylphosphine ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Methyl phosphine

15-6500-5G 15-6500-25G
100209-740EA 193.91 USD
100209-740 100209-320
Trimethylphosphine ≥98%
Trimethylphosphine

Phosphine

Formula: C₃H₉P
MW: 76.08 g/mol
Boiling Pt: 37.8 °C (1013 hPa)
Melting Pt: -85 °C
Density: 0.748 g/cm³ (10 °C)
Flash Pt: –22 °F
MDL Number: MFCD00008510
CAS Number: 594-09-2
EINECS: 209-823-1
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR