Triethyl phosphite ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

15-6350-1000G 15-6350-250G
100206-420EA 147.2 USD
100206-420 100206-422
Triethyl phosphite ≥98%
Triethyl phosphite
Formula: C₆H₁₅O₃P
MW: 166.16 g/mol
Boiling Pt: 156 °C (1013 hPa)
Melting Pt: 51 °C
Density: 0.9643 g/cm³ (20 °C)
Flash Pt: 52 °C
MDL Number: MFCD00009084
CAS Number: 122-52-1
EINECS: 204-552-5
UN: 2323
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR