Tris(4-chlorophenyl)phosphine ≥99%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Tri(p-chlorophenyl)phosphine, Tris(p-chlorophenyl)phosphine
15-6050-1G
15-6050-5G
100204-524EA
127.38
USD
100204-524
100206-092
Tris(4-chlorophenyl)phosphine ≥99%
Tris(4-chlorophenyl)phosphine
Phosphine
Formula:
C₁₈H₁₂Cl₃P MW: 365.63 g/mol Boiling Pt: 431 °C (1013 hPa) Melting Pt: 90…93 °C Density: 1.449 g/cm³ (20 °C) |
MDL Number:
MFCD00013639 CAS Number: 1159-54-2 EINECS: 214-596-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...