Tris(3-chlorophenyl)phosphine ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Tri(m-chlorophenyl)phosphine, Tris(m-chlorophenyl)phosphine

15-6030-5G 15-6030-1G
100206-090EA 400.55 USD
100206-090 100200-432
Tris(3-chlorophenyl)phosphine ≥97%
Tris(3-chlorophenyl)phosphine

Phosphine

Formula: C₁₈H₁₂Cl₃P
MW: 365.63 g/mol
Melting Pt: 64…66 °C
MDL Number: MFCD00013632
CAS Number: 29949-85-7
EINECS: 249-970-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR