Tributylphosphine 10% (w/w) in hexanes ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Tri-n-butylphosphine, TBP

15-5802-250G 15-5802-1000G
100211-022EA 172.68 USD
100211-022 100207-590
Tributylphosphine 10% (w/w) in hexanes ≥99%
Tributylphosphine

Phosphine

Formula: C₁₂H₂₇P
MW: 202.32 g/mol
Boiling Pt: 240 °C (1013 hPa)
Melting Pt: –65 °C
Density: 0.8165 g/cm³ (20 °C)
Flash Pt: 40 °C
MDL Number: MFCD00009462
CAS Number: 998-40-3
EINECS: 213-651-2
UN: 1993
ADR: 3,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR