Tributylphosphine ≥93%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Tri-n-butylphosphine, TBP
15-5801-500G
15-5801-100G
100208-840EA
315.63
USD
100208-840
100206-404
Tributylphosphine ≥93%
Tributylphosphine
Phosphine
Formula:
C₁₂H₂₇P MW: 202.32 g/mol Boiling Pt: 110…115 °C (10…12 mmHg) Melting Pt: –65 °C Density: 0.8165 g/cm³ (20 °C) Flash Pt: 99 °F |
MDL Number:
MFCD00009462 CAS Number: 998-40-3 EINECS: 213-651-2 UN: 2845 ADR: 4.2,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...