Tributylphosphine ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Tri-n-butylphosphine, TBP

15-5800-500G 15-5800-100G
100209-314EA 1252.62 USD
100209-314 100209-312
Tributylphosphine ≥99%
Tributylphosphine

Phosphine

Formula: C₁₂H₂₇P
MW: 202.32 g/mol
Boiling Pt: 240
Melting Pt: -65
Density: 0.817
Flash Pt: 99 °F
MDL Number: MFCD00009462
CAS Number: 998-40-3
EINECS: 213-651-2
UN: 2845
ADR: 4.2 I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR