Dichloro(phenyl)phosphine ≥99%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
P,P-Dichlorophenylphosphine, Phenyldichlorophosphine
15-4155-250G
15-4155-50G
100206-088EA
786.95
USD
100206-088
100204-522
Dichloro(phenyl)phosphine ≥99%
Dichloro(phenyl)phosphine
Phosphine
Formula:
C₆H₅Cl₂P MW: 178.98 g/mol Boiling Pt: 222 °C (1013 hPa) Melting Pt: -51 °C Density: 1.32 g/cm³ (20 °C) Flash Pt: >230 °F Storage Temperature: Ambient |
MDL Number:
MFCD00000528 CAS Number: 644-97-3 EINECS: 211-425-8 UN: 2798 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...