Methyldiphenylphosphine ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

15-3250-25G 15-3250-100G 15-3250-5G
100201-484EA 301.48 USD
100201-484 100197-412 100206-386
Methyldiphenylphosphine ≥99%
Methyldiphenylphosphine

Phosphine

Formula: C₁₃H₁₃P
Boiling Pt: 118…120 °C (2.2 mmHg)
Melting Pt: 28 °C
Density: 1.0784 g/cm³ (15 °C)
Flash Pt: >230 °F
MDL Number: MFCD00008508
CAS Number: 1486-28-8
EINECS: 216-065-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR