(Ethyl)triphenylphosphonium bromide ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Ethyltriphenylphosphonium bromide

15-2301-25G 15-2301-100G
100204-952EA 82.09 USD
100204-952 100200-646
(Ethyl)triphenylphosphonium bromide ≥99%
(Ethyl)triphenylphosphonium bromide
Formula: C₂₀H₂₀BrP
MW: 371.26 g/mol
Melting Pt: 206…208 °C
Storage Temperature: Ambient
MDL Number: MFCD00011838
CAS Number: 1530-32-1
EINECS: 216-223-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR