Diphenylphosphine 10% (w/w) in hexanes ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

15-1702-500G 15-1702-100G
100199-794EA 444.43 USD
100199-794 100207-578
Diphenylphosphine 10% (w/w) in hexanes ≥99%
Diphenylphosphine

Phosphine

Formula: C₁₂H₁₁P
MW: 186.19 g/mol
Density: 0.68
MDL Number: MFCD00003040
CAS Number: 829-85-6
EINECS: 212-591-4
UN: 1993
ADR: 3,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR