Diphenylphosphine ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

15-1700-10G 15-1700-50G
100206-360EA 97.66 USD
100206-360 100206-362
Diphenylphosphine ≥99%
Diphenylphosphine

Phosphine

Formula: C₁₂H₁₁P
MW: 186.19 g/mol
Boiling Pt: 116…119 °C (2,6 mmHg)
Density: 1,07
MDL Number: MFCD00003040
CAS Number: 829-85-6
EINECS: 212-591-4
UN: 2845
ADR: 4.2,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR