Chlorodiphenylphosphine ≥95%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: DPPC, Diphenylchlorophosphine

15-1551-50G 15-1551-250G 15-1551-1000G
100204-950EA 80.68 USD
100204-950 100204-948 100202-366
Chlorodiphenylphosphine ≥95%
Chlorodiphenylphosphine

Phosphine

Formula: C₁₂H₁₀ClP
MW: 220.64 g/mol
Boiling Pt: 100…102 °C (1 mmHg)
Melting Pt: 14…16 °C
Density: 1.2 g/cm³ (20 °C)
Flash Pt: >230 °F
Storage Temperature: Ambient
MDL Number: MFCD00000529
CAS Number: 1079-66-9
EINECS: 214-093-2
UN: 1760
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR