1,2-Bis(diphenylphosphino)ethane ≥99%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Ethylenebis(diphenylphosphine), Bis(1,2-diphenylphosphino)ethane, DIPHOS, 1,2-Bis(diphenylphosphino)ethane, Ethane-1,3-diylbis(diphenylphosphine)
15-0200-100G
15-0200-5G
15-0200-25G
100198-126EA
543.51
USD
100198-126
100197-392
100200-416
1,2-Bis(diphenylphosphino)ethane ≥99%
1,2-Bis(diphenylphosphino)ethane
Phosphine
Formula:
C₂₆H₂₄P₂ MW: 398.42 g/mol Melting Pt: 140 - 142 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003047 CAS Number: 1663-45-2 EINECS: 216-769-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...