1,4-Bis(diphenylphosphino)butane ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
DPPB, 1,4-Bis(diphenylphosphino)butane, Butylenebis(diphenylphosphine), Bis(1,2-diphenylphosphino)butane, Butane-1,3-diylbis(diphenylphosphine)
15-0170-10G
15-0170-50G
15-0170-2G
100202-954EA
171.26
USD
100202-954
100200-854
100198-608
1,4-Bis(diphenylphosphino)butane ≥98%
1,4-Bis(diphenylphosphino)butane
Phosphine
Formula:
C₂₈H₂₈P₂ Melting Pt: 132…134 °C Density: 1.23 g/cm³ (20 °C) |
MDL Number:
MFCD00003051 CAS Number: 7688-25-7 EINECS: 231-698-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...