(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (R)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, (R)-(+)-BINAP, (R)-BINAP, (R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthalene, (R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl

15-0150-5G 15-0150-1G 15-0150-250MG
100210-998EA 407.63 USD
100210-998 100210-996 100198-600
(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl ≥98%
(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl

BINAP, Phosphine

Formula: C₄₄H₃₂P₂
Melting Pt: 240.5…242 °C
MDL Number: MFCD00010805
CAS Number: 76189-55-4
Merck Index: 13,01223

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR