1,2-Bis(dimethylphosphino)ethane ≥98%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Ethane-1,3-diylbis(dimethylphosphine), 1,2-Bis(dimethylphosphino)ethane, Bis(1,2-dimethylphosphino)ethane, DMPE, Ethylenebis(dimethylphosphine)
15-0090-1G
15-0090-5G
100210-992EA
261.85
USD
100210-992
100210-994
1,2-Bis(dimethylphosphino)ethane ≥98%
1,2-Bis(dimethylphosphino)ethane
Phosphine
Formula:
C₆H₁₆P₂ MW: 150.14 g/mol Boiling Pt: 180 °C Melting Pt: 147…149 °C Density: 1.1 g/cm³ (20 °C) Flash Pt: 2 °F |
MDL Number:
MFCD00008511 CAS Number: 23936-60-9 UN: 1993 ADR: 3,II |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...