(Trifluoromethyl)trimethylsilane ≥97%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
trimethyl(trifluoromethyl)silane
14-7990-25G
14-7990-5G
100210-986EA
503.88
USD
100210-986
100210-988
(Trifluoromethyl)trimethylsilane ≥97%
(Trifluoromethyl)trimethylsilane
Formula:
C₄H₉F₃Si MW: 142.2 g/mol Boiling Pt: 55.5 °C (1013 hPa) Density: 0.95 g/cm³ (20 °C) Flash Pt: 14 °F |
MDL Number:
MFCD00145454 CAS Number: 81290-20-2 UN: 1993 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...