Triethylsilylacetylene ≥97%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
14-7850-5G
14-7850-25G
100206-300EA
151.45
USD
100206-300
100206-298
Triethylsilylacetylene ≥97%
Triethylsilylacetylene
Formula:
C₈H₁₆Si MW: 140.3 g/mol Density: 0,783 Flash Pt: 63 °F |
MDL Number:
MFCD00075373 CAS Number: 1777-03-3 UN: 1993 ADR: 3,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...