Silicon 99,9999, chips
Supplier: STREM CHEMICALS INC MS
|
Warning
|
14-0600-500G
14-0600-100G
100209-650EA
873.29
USD
100209-650
100196-040
Silicon 99,9999, chips
Silicon
Formula:
Si MW: 28.09 g/mol Boiling Pt: 2600 °C Melting Pt: 1420 °C Density: 2.42 Storage Temperature: Ambient |
MDL Number:
MFCD00085311 CAS Number: 7440-21-3 EINECS: 231-130-8 Merck Index: 12,08635 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...