Aluminium oxide, γ-phase low sodium

Supplier: STREM CHEMICALS INC MS
Warning

13-2610-2000G 13-2610-500G
100199-756EA 554.83 USD
100199-756 100206-294
Aluminium oxide, γ-phase low sodium
Aluminium oxide, γ-phase
Formula: Al₂O₃
MW: 101.96 g/mol
Boiling Pt: 2980 °C (1013 hPa)
Melting Pt: 2045 °C
Density: 3.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR