Aluminium nitride, agglomerated powder
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Aluminum nitride
13-1377-100G
13-1377-500G
100208-798EA
106.15
USD
100208-798
100203-728
Aluminium nitride, agglomerated powder
Aluminium nitride
Formula:
AlN Boiling Pt: 2517 °C (1013 hPa) Melting Pt: 1900 - 2200 °C Density: 3.257 to 3.267 g/cm³ (160 °C) |
MDL Number:
MFCD00003429 CAS Number: 24304-00-5 EINECS: 246-140-8 Merck Index: 12,00366 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...