α-Alumina trihydrate 65% Al2O3, 34.8% H2O, 0.15% Na2O

Supplier: STREM CHEMICALS INC MS
Warning

13-1000-500G 13-1000-2000G
100199-754EA 72.18 USD
100199-754 100198-116
α-Alumina trihydrate 65% Al2O3, 34.8% H2O, 0.15% Na2O
α-Alumina trihydrate
Formula: Al₂O₃·3H₂O
MDL Number: MFCD00149140
CAS Number: 1344-28-1
EINECS: 215-691-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR