Aluminium fluoride, anhydrous ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Aluminum fluoride

13-0075-10G 13-0075-50G
100199-350EA 124.55 USD
100199-350 100202-922
Aluminium fluoride, anhydrous ≥99%
Aluminium fluoride
Formula: AlF₃
MW: 83.98 g/mol
Melting Pt: 1040 (subl.)
Density: 2.88 g/cm³ (20 °C)
MDL Number: MFCD00003426
CAS Number: 7784-18-1
EINECS: 232-051-1
UN: 3260
ADR: 8 II
Merck Index: 12,00351

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR