Aluminium chloride, anhydrous ≥99.99% (Al basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Aluminum chloride

13-0050-250G 13-0050-50G
100201-430EA 953.97 USD
100201-430 100199-752
Aluminium chloride, anhydrous ≥99.99% (Al basis), PURATREM
Aluminium chloride
Formula: AlCl₃
MW: 133.34 g/mol
Boiling Pt: 180 °C (subl.)
Melting Pt: 190 °C
Density: liquid 1.31 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003422
CAS Number: 7446-70-0
EINECS: 231-208-1
UN: 1726
ADR: 8,II
Merck Index: 13,00335

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR