Sodium cyanotrihydroborate ≥95%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Sodium cyanoborohydride
11-2800-5G
11-2800-25G
100209-636EA
55.2
USD
100209-636
100202-918
Sodium cyanotrihydroborate ≥95%
Sodium cyanotrihydroborate
Formula:
CH₃BNNa MW: 62.84 g/mol Melting Pt: > 242 °C (dec.) Density: 1.11 to 1.12 g/cm³ (28 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003516 CAS Number: 25895-60-7 EINECS: 247-317-2 UN: 3134 ADR: 4.3,I Merck Index: 13,08680 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...