Sodium fluoride ≥99.99% (Na basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

11-2000-100G 11-2000-25G
100202-364EA 525.11 USD
100202-364 100203-720
Sodium fluoride ≥99.99% (Na basis), PURATREM
Sodium fluoride
Formula: NaF
MW: 41.99 g/mol
Boiling Pt: 1700 °C (1013 hPa)
Melting Pt: 993 °C
Density: 2.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003524
CAS Number: 7681-49-4
EINECS: 231-667-8
UN: 1690
ADR: 6.1,III
Merck Index: 13,08691
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2807d129a60e4d136eb782386d584317338c4499&vid=02396e9a739aa8e35f0b20961710e28c6ec76be0&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR