Anti-Salmonella CSA-1 Antibody

Supplier: SeraCare
5310-0322
95058-076EA 1041.62 USD
95058-076
Anti-Salmonella CSA-1 Antibody
Antibodies
Primary Goat Anti-Salmonella CSA-1 Reacts with Salmonella

Type: Primary
Antigen: Salmonella CSA-1
Clonality: Polyclonal
Clone:
Conjugation: (none)
Epitope:
Host: Goat
Isotype:
Reactivity: Salmonella
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR