Anti-IgG gamma GoatPolyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: SeraCare
5230-0305
95058-136EA 230.6 USD
95058-136
Anti-IgG gamma GoatPolyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Secondary Goat Anti-IgG (g) Reacts with Mouse

Type: Secondary
Antigen: IgG gamma
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope: gamma chain
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR