Anti-GFP Chicken Polyclonal Antibody

Supplier: Rockland Immunochemical
600-901-215 600-901-B12
RL600-901-215EA 405.37 USD
RL600-901-215 RL600-901-B12
Anti-GFP Chicken Polyclonal Antibody
Antibodies
Polyclonal anti-GFP is designed to detect GFP and its variants. This antibody can be used to detect GFP by ELISA (sandwich or capture) for the direct binding of antigen and recognizes wild type, recombinant and enhanced forms of GFP.

Recommended Dilutions: ELISA: 1:2000 - 1:12000; Western Blot: 1:200 - 1:1000; Immunochemistry: 1:200 - 1:1000;

Type: Tag
Antigen: GFP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Chicken
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR