Fmoc-Pro-OH

Supplier: Chem-Impex International

02448-100G 02448-1KG 02448-25G
CH02448-100GEA 75 USD
CH02448-100G CH02448-1KG CH02448-25G
Fmoc-Pro-OH
Fmoc-Pro-OH
Formula: C₂₀H₁₉NO₄
MDL Number: MFCD00037122
CAS Number: 71989-31-6
EINECS: 276-259-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR