Edge® 40-105 Fully Coated Nitrile Gloves, Ansell
Supplier: Ansell Healthcare
218902 218904 218900 218903
32890-194CS 1285.5 USD
32890-194 32890-198 32890-190 32890-196
Edge® 40-105 Fully Coated Nitrile Gloves, Ansell
Gloves
Fully coated nitrile gloves outwear standard 8-ounce cotton gloves by 10-to-1 and provide superior snag, puncture, abrasion, and cut resistance.
  • Oil channeled away by porous coating
  • Ergonomically designed for greater comfort
  • Excellent dexterity
The porous nitrile coating channels oil away from glove surface for excellent grip in oily applications and molds to the hand for greater dexterity.
Use in the appliance manufacturing, automotive and transportation, general handling, and metal fabrication industries. Applications include assembling, inspecting and packing small or delicate parts; engine and transmission assembly; general maintenance and plant use; general material handling; and shipping and receiving/handling cartons and packaging.
Ergonomically designed with no uncomfortable seams.
Certificaciones: ANSI level 1 cut protection. ANSI level 4 abrasion protection.
Envase: Gloves are packaged 12 pairs in a bag; 12 bags in a carton.