MAS™ UrichemTRAK™ Controls, Thermo Scientific
Supplier: MICROGENICS CORPORATION (MAS)
UR11001 UR-MP UR22002 10014127
89130-682EA 351.1 USD
89130-682 89130-686 89130-684 10154-288
MAS™ UrichemTRAK™ Controls, Thermo Scientific
Controls and Calibration Verification Materials
Monitor the most commonly tested urine chemistry analytes with liquid, assayed MAS™ UrichemTRAK™ Controls, available in a convenient Vista™ vial format
Liquid, human urine matrix. Open and use, no equilibration required. Two-year open-vial stability. Individual levels or multi-pack configurations. Stable for 24 months at 2 to 8°C. Assayed values available for most major analyzers.
Analytes: Amylase, Calcium, Chloride, Cortisol, Creatinine, Glucose, Magnesium, Microalbumin, Phosphorus, Potassium, Sodium, Total Protein, Urea Nitrogen, Uric Acid.
Ready-to-use and requiring no additional preparation, these controls will not interfere with ISEs.
Información para pedidos: Available in Vista™ vial format.