MAS™ UA Controls, Thermo Scientific
Supplier: MICROGENICS CORPORATION (MAS)
Monitor urine dipstick and microscopic components with MAS™ UA Controls, bi-level controls with improved room temperature storage capability
Available in convenient 15mL dropper bottles.
Analytes: Bilirubin, Blood, Creatinine, Crystals, Glucose, hCG, Ketones, Leukocyte Esterase, Microalbumin, Nitrite, pH, Potassium, Protein, Red Blood Cells, Sodium, Specific Gravity, Urobilinogen, White Blood Cells.
Liquid and ready-to-use, this urine control is assayed for most major reagents strips and strip systems. It includes 10 commonly-assayed dipstick tests plus Microalbumin, Creatinine, and hCG.
Información para pedidos: Control designed to monitor urine dipstick and microscopic components. Available in individual levels only.