Thermo Scientific™ MAS™ Urinalysis Controls, Thermo Scientific
Supplier: MICROGENICS CORPORATION (MAS)
024222
024225
024227
024224
89130-662EA
133.39
USD
89130-662
89130-666
89130-668
89130-664
Thermo Scientific™ MAS™ Urinalysis Controls, Thermo Scientific
Controls and Calibration Verification Materials
Combine dipstick methods and microscopic components with bi-level, liquid assayed MAS™ Urinalysis Controls
Human urine matrix.
Analytes: Bilirubin, Blood, Creatinine, Crystals, Glucose, hCG, Ketones, Leukocyte Esterase, Microalbumin, Nitrite, Osmolality, pH, Potassium, Protein, Red Blood Cells, Sodium, Specific Gravity, Urobilinogen, White Blood Cells.
This control is assayed for most major reagent strips and strip systems.