Methacrylic acid 99.5% stabilized, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Methylpropenoic acid

AC16831-0250 AC16831-0010
200058-020EA 47.8 USD
200058-020 200058-010 200058-016 200058-012 103526-220
Methacrylic acid 99.5% stabilized, Extra Pure
Methacrylic acid
Formula: C₄H₆O₂
MW: 86.09 g/mol
Boiling Pt: 163 °C (760 mmHg)
Melting Pt: 16 °C
Density: 1.02 g/cm³ (20 °C)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00002651
CAS Number: 79-41-4
EINECS: 201-204-4
UN: 2531
ADR: 8,II
Merck Index: 13,05967

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR