4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane (Kryptofix® 222) ≥99% (by GC) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.10647.0005 8.10647.0001 8.10647.8250
EM8.10647.0005EA 723 USD
EM8.10647.0005 EM8.10647.0001 EM8.10647.8250
4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane (Kryptofix® 222) ≥99% (by GC) for synthesis, Sigma-Aldrich®
4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane (Kryptofix® 222)
Formula: C₁₈H₃₆N₂O₆
Melting Pt: 69…71 °C
Storage Temperature: Ambient
MDL Number: MFCD00005111
CAS Number: 23978-09-8
EINECS: 245-962-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a74ac532561e4fecf111f3260b402bcd1fb0af9&vid=13491901da172b5971e6d7f330c893460a77cd23&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (GC) ≥ 99
Identity (IR-spectrum) passes test
Melting range (°C) 70 - 73

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR