Methylammonium chloride, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: MACl, Methylammonium chloride, Aminomethane hydrochloride, Methylamine hydrochloride

M-Clarity™ quality level = MQ200

8.06020.1000 8.06020.9025
EM8.06020.1000EA 201 USD
EM8.06020.1000 EM8.06020.9025
Methylammonium chloride, Sigma-Aldrich®
Methylammonium chloride
Formula: CH₆ClN
MDL Number: MFCD00012849
CAS Number: 593-51-1
EINECS: 209-795-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR