4-Bromo-2,2-difluoro-1,3-benzodioxole 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24389-03
B24389-06
AAB24389-03EA
62.6
USD
AAB24389-03
AAB24389-06
4-Bromo-2,2-difluoro-1,3-benzodioxole 97%
4-Bromo-2,2-difluoro-1,3-benzodioxole
Purity: 97%
CAS No.: 144584-66-7
Molecular Formula: C7H3BrF2O2
Formula Weight: 237.00
Refractive Index: 1.4968
Formula:
C₇H₃BrF₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD01631385 CAS Number: 144584-66-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...