3,4-Dihydroxybenzoic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Protocatechuic acid

B24016-14 B24016-22 B24016-36
AAB24016-14EA 85.48 USD
AAB24016-14 AAB24016-22 AAB24016-36
3,4-Dihydroxybenzoic acid 97%
3,4-Dihydroxybenzoic acid

Purity: 97% CAS No.: 99-50-3 Molecular Formula: C7H6O4 Formula Weight: 154.12 Melting Point: ca 202° dec. Beilstein Reference: 1448841 Merck Reference: 14,7894 MDL No.: MFCD00002509

Formula: C₇H₆O₄
MW: 154.12 g/mol
Boiling Pt: 125 - 135 °C (0.001 -rr)
Melting Pt: ca 202 °C (dec.)
Density: 1.5415 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002509
CAS Number: 99-50-3
EINECS: 202-760-0
Merck Index: 12,08080

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR