Benzyltributylammonium bromide 99%

Supplier: Thermo Scientific Chemicals
Warning

B23941-18 B23941-14 B23941-30
AAB23941-18EA 63.4 USD
AAB23941-18 AAAB23941-14 AAB23941-30
Benzyltributylammonium bromide 99%
Benzyltributylammonium bromide

Purity: 99% CAS No.: 25316-59-0 Molecular Formula: [CH3(CH2)3]3N(CH2C6H5)Br Formula Weight: 356.40 Melting Point: ca 175° dec. Beilstein Reference: 3776294

Formula: C₁₉H₃₄BrN
MW: 356.39 g/mol
Melting Pt: ∼ 175 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011848
CAS Number: 25316-59-0
EINECS: 246-819-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR