N-Phenyl-p-phenylenediamine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Aminodiphenylamine, N-(4-Aminophenyl)aniline
B23867-30
B23867-0B
B23867-22
AAB23867-30EA
69.9
USD
AAB23867-30
AAB23867-0B
AAAB23867-22
N-Phenyl-p-phenylenediamine 98%
N-Phenyl-p-phenylenediamine
Purity: 98%
CAS No.: 101-54-2
Molecular Formula: C12H12N2
Formula Weight: 184.24
Flash Point: 193°(379°F)
Melting Point: 72-76°
Boiling Point: 354°
Beilstein Reference: 908935
UN No.: 3077
MDL No.: MFCD00007850
Formula:
C₁₂H₁₂N₂ MW: 184.24 g/mol Boiling Pt: 354 °C Melting Pt: 72…76 °C Density: 1.15 g/cm³ (20 °C) Flash Pt: 193 °C (379 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007850 CAS Number: 101-54-2 EINECS: 202-951-9 UN: 3077 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...