5-Bromo-2,3-difluorophenol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Bromo-3,4-difluoro-5-hydroxybenzene
B23853-03
B23853-14
AAB23853-03EA
27.6
USD
AAB23853-03
AAAB23853-14
AAB23853-06
5-Bromo-2,3-difluorophenol 97%
5-Bromo-2,3-difluorophenol
Purity: 97%
CAS No.: 186590-26-1
Molecular Formula: C6H3BrF2O
Formula Weight: 208.99
Refractive Index: 1.5380
Beilstein Reference: 8831044
Formula:
C₆H₃BrF₂O MW: 208.99 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01631350 CAS Number: 186590-26-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...