2-Amino-5-nitrobenzophenone 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (2-Amino-5-nitrophenyl)(phenyl)methanone

B23773-14 B23773-22 B23773-06
AAB23773-14EA 60 USD
AAB23773-14 AAB23773-22 AAB23773-06
2-Amino-5-nitrobenzophenone 98%
2-Amino-5-nitrobenzophenone

Purity: 98% CAS No.: 1775-95-7 Molecular Formula: C13H10N2O3 Formula Weight: 242.23 Melting Point: 166-168° MDL No.: MFCD00007364

Formula: C₁₃H₁₀N₂O₃
MW: 242.23 g/mol
Melting Pt: 163.5 °C
Density: 1.474 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007364
CAS Number: 1775-95-7
EINECS: 217-207-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR