4-Ethoxyphenylboronic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Ethoxybenzeneboronic acid, (4-Ethoxyphenyl)metaboric acid, Dihydroxy(4-ethoxyphenyl)borane

B23683-14 B23683-06 B23683-03
AAB23683-14EA 283 USD
AAB23683-14 AAB23683-06 AAB23683-03
4-Ethoxyphenylboronic acid 98%
4-Ethoxyphenylboronic acid

Purity: 98% CAS No.: 22237-13-4 Molecular Formula: C8H11BO3 Formula Weight: 165.99 Melting Point: 165-168° Beilstein Reference: 3530408

Formula: C₈H₁₁BO₃
MW: 165.98 g/mol
Melting Pt: 165…168 °C
Storage Temperature: Ambient
MDL Number: MFCD00674028
CAS Number: 22237-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR