3-(Aminomethyl)pyridine 98+%

Supplier: Thermo Scientific Chemicals
Danger

B23634-14 B23634-22
AAB23634-14EA 45.6 USD
AAB23634-14 AAB23634-22
3-(Aminomethyl)pyridine 98+%
3-(Aminomethyl)pyridine

Purity: 98+% CAS No.: 3731-52-0 Molecular Formula: C6H8N2 Formula Weight: 108.14 Flash Point: 100°(212°F) Refractive Index: 1.5510 Odor: Amine-like pH: 11-12 Melting Point: -21° Boiling Point: 226° Beilstein Reference: 108056 UN No.: 2735 MDL No.: MFCD00006412

Formula: C₆H₈N₂
Boiling Pt: 226 °C
Melting Pt: –21 °C
Density: 1.062
Flash Pt: 100 °C (212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006412
CAS Number: 3731-52-0
EINECS: 223-091-0
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR